missing translation for 'onlineSavingsMsg'
Learn More

Azadibenzocyclooctyne acid

Product Code. p-7042950
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15486449 25 mg 25mg
15496449 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15486449 Supplier Thermo Scientific Alfa Aesar Supplier No. J64549.MB
Add to basket
This item is not returnable. View return policy

CAS: 1425485-72-8 | C21H19NO3 | 333.39 g/mol

Azadibenzocyclooctyne acid is the simplest amine reactive reagent for introducing a dibenzylcyclooctyne to available amine functionality.Azadibenzocyclooctyne acid is widely useful in strain-promoted copper-free azide-alkyne click chemistry reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azadibenzocyclooctyne acid is the simplest amine reactive reagent for introducing a dibenzylcyclooctyne to available amine functionality.Azadibenzocyclooctyne acid is widely useful in strain-promoted copper-free azide-alkyne click chemistry reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.

Solubility
Soluble in dimethyl sulfoxide and dimethyl formamide.

Notes
Store in cool place. Incompatible with amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1425485-72-8
Molecular Formula C21H19NO3
Molecular Weight (g/mol) 333.39
MDL Number MFCD22380737
InChI Key NIRLBCOFKPVQLM-UHFFFAOYSA-N
Synonym dbco-c6-acid, azadibenzocyclooctyne acid, dibenzocyclooctyne-acid, storage temp.:-20c, 6-11,12-didehydro-5,6-dihydrodibenzo b,f azocine-5-yl-6-oxohexanoic acid
PubChem CID 77078156

Specifications

Color White to Gray
Quantity 25 mg
Solubility Information Soluble in dimethyl sulfoxide and dimethyl formamide.
Formula Weight 333.38
Physical Form Solid
Chemical Name or Material Azadibenzocyclooctyne acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.