accessibility menu, dialog, popup

UserName

NOOOH

CAS RN 1425485-72-8

IUPAC Name: 6-{2-azatricyclo[10.4.0.0⁴,⁹]hexadeca-1(16),4,6,8,12,14-hexaen-10-yn-2-yl}-6-oxohexanoic acid
Synonyms: 6-{2-azatricyclo[10.4.0.0⁴,⁹]hexadeca-1(16),4,6,8,12,14-hexaen-10-yn-2-yl}-6-oxohexanoic acid
Molecular Weight (g/mol): 333.39
Molecular Formula: C21H19NO3
InChi Key: NIRLBCOFKPVQLM-UHFFFAOYSA-N
SMILES: OC(=O)CCCCC(=O)N1CC2=CC=CC=C2C#CC2=CC=CC=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results