Includes
- 1.NB100-2220: LC3B/MAP1LC3B Antibody
- 2.NB110-53818: ATG5 Antibody
- 3.NB110-56893: ATG9A Antibody
- 4.NB110-60928: ATG16L1 Antibody
- 5.NB110-87318: Beclin 1/ATG6 Antibody
Autophagy specifically detects Autophagy in Alligator, Bovine, Canine, Human, Mouse, Pig, Primate, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Simple Western, Western Blot.
Specifications
Specifications
| Applications | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, KnockDown, Simple Western, Western Blot |
| Target Species | Alligator, Bovine, Canine, Human, Mouse, Pig, Primate, Rat |