missing translation for 'onlineSavingsMsg'
Learn More

Aspartate Aminotransferase Antibody, Novus Biologicals™

Product Code. 18214790 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214790 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214790 Supplier Novus Biologicals Supplier No. NB10093403
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Goat Polyclonal Antibody

Aspartate Aminotransferase Polyclonal specifically detects Aspartate Aminotransferase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aspartate Aminotransferase
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.01 - 1 ug/ml, Immunohistochemistry 2 - 5 ug/ml, Immunohistochemistry-Paraffin 2 - 5 ug/ml, Peptide ELISA Detection limit 1:4000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_002070.1
Gene Alias aspartate aminotransferase, cytoplasmic, EC 2.6.1.1, GIG18, Glutamate oxaloacetate transaminase 1, glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1), growth-inhibiting protein 18, Transaminase A
Gene Symbols GOT1
Host Species Goat
Immunogen Peptide with sequence C-RSYRYWDAEKR corresponding to internal region according to NP_002070.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2805
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.