Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human ASNS. The Asparagine synthetase Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 440 |
| Product Line | Novus |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | asparagine synthetase, asparagine synthetase (glutamine-hydrolyzing), asparagine synthetase [glutamine-hydrolyzing], Cell cycle control protein TS11, EC 6.3.5.4, Glutamine-dependent asparagine synthetase, TS11, TS11 cell cycle control protein |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Symbol | ASNS |
| Molecular Weight (g/mol) | 32 kDa |
| Quantity | 100 μL |
| Source | E. coli |
| Specific Reactivity | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54142. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |