Learn More
Invitrogen™ Antibody Diluent
Product Code. 10414753
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
500mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10414753 250 mL Each
15908982 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10414753 Supplier Invitrogen™ Supplier No. 003118
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Antibody Diluent
Ready-to-use diluent with BSA (for stabilizing the antibody) and preservative.
“This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.”
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ready-to-Use Antibody Diluent
For Use With (Application) Cell analysis, cellular imaging, immunohistochemistry (IHC), IHC staining and detection
Physical Form Liquid
Quantity 250 mL
Includes Antibody Diluent, 250mL, Liquid, Ready-to-Use Diluent, 26628-22-8 CAS Number, 0.1% Sodium Azide Preservative, BSA Stabilizer Additives, Highly Purified Grade, Wet Ice Shipping Condition, 2 to 8°C Storage Temperature, For In-vitro Diagnostic Use
For In Vitro Diagnostic Use.