missing translation for 'onlineSavingsMsg'
Learn More

Anthracene-d10, TRC

Product Code. 30179037
Change view
Click to view available options
Quantity:
100 mg
1 g
2.5 g
Unit Size:
100mg
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179037 100 mg 100mg
30179063 1 g 1g
30179330 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179037 Supplier Toronto Research Chemicals Supplier No. TRCA678403100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1719-06-8
Molecular Formula C14 2H10
Molecular Weight (g/mol) 188.29
Synonym Anthracene D10, Anthracene-d10 (7CI,8CI,9CI), Perdeuterated anthracene, Perdeuterioanthracene, Perdeuteroanthracene
IUPAC Name 1,2,3,4,5,6,7,8,9,10-decadeuterioanthracene
SMILES [2H]c1c([2H])c([2H])c2c([2H])c3c([2H])c([2H])c([2H])c([2H])c3c([2H])c2c1[2H]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H10/c1-2-6-12-10-14-8-4-3-7-13(14)9-11(12)5-1/h1-10H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
Formula Weight 188.141
Chemical Name or Material Anthracene D10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.