CAS RN 1719-06-8
IUPAC Name:
(²H₁₀)anthracene
Synonyms:
(²H₁₀)anthracene
Molecular Weight (g/mol):
188.3
Molecular Formula:
C14H10
InChi Key:
MWPLVEDNUUSJAV-LHNTUAQVSA-N
SMILES:
[2H]C1=C([2H])C2=C([2H])C3=C([2H])C([2H])=C([2H])C([2H])=C3C([2H])=C2C([2H])=C1[2H]
1
–
2
of
2
results
1
Anthracene-d10, TRC
CAS: 1719-06-8 Molecular Formula: C14 2H10 Molecular Weight (g/mol): 188.29 Synonym: Anthracene D10,Anthracene-d10 (7CI,8CI,9CI),Perdeuterated anthracene,Perdeuterioanthracene,Perdeuteroanthracene IUPAC Name: 1,2,3,4,5,6,7,8,9,10-decadeuterioanthracene SMILES: [2H]c1c([2H])c([2H])c2c([2H])c3c([2H])c([2H])c([2H])c([2H])c3c([2H])c2c1[2H]
2
Anthracene-d10, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.