Learn More
Ammonium Nitrate, Extra Pure, MilliporeSigma™
Product Code. 15886960
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15886960 5 kg Plastic Bottle 5kg
15876960 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15886960 Supplier Supelco Supplier No. 1011875000
 Restricted Item
Estimated Shipment: 26-11-2026
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
CAS: 6484-52-2 | Formula: H4N2O3 | Weight: 80.04 g/mol
Recommended Storage
5°C to +30°C
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O
Specifications
Boiling Point 210°C (15.0hPa)
Color Colorless
pH 4.5 to 7.0 (at 100g/L 20°C)
Physical Form Solid
Quantity 5 kg
Packaging Plastic Bottle
Vapor Pressure 125 at 20°C
Density 1.72g/3
Solubility Information 1,920g/L at 20°C
Formula Weight 80.04
Percent Purity >95.0%
Grade Extra Pure