accessibility menu, dialog, popup

UserName

ONOONH4

CAS RN 6484-52-2

IUPAC Name: ammonium nitrate
Synonyms: ammonium nitrate ammonium(I) nitrate (1:1) nitrate of ammonia nitrate d'ammonium nitric acid, ammonium salt nitric acid ammonium salt (1:1) ammonium saltpeter ammonium nitricum nitrato amonico
Molecular Weight (g/mol): 80.04
Molecular Formula: H4N2O3
InChi Key: DVARTQFDIMZBAA-UHFFFAOYSA-O
SMILES: [NH4+].[O-][N+]([O-])=O
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
Physical Form 
  • (1)
  • (5)
  • (1)
  • (2)
  • (2)
  • (6)
Grade 
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (7)
  • (1)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (2)
  • (2)
  • (1)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (2)
  • (6)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (7)
  • (1)
  • (4)