missing translation for 'onlineSavingsMsg'
Learn More

Amiodarone, TRC

Product Code. 17917754
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17917754 10 g 10g
17979564 1 g 1g
17929474 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17917754 Supplier Toronto Research Chemicals Supplier No. TRCA63294510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1951-25-3
Molecular Formula C25H29I2NO3
Molecular Weight (g/mol) 645.31
Synonym 2-butyl-3-benzofuranyl 4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl Ketone, 2-Butyl-3-[3,5-diiodo-4-(2-diethylaminoethoxy)benzoyl]benzofuran, 2-Butyl-3-benzofuranyl p-[(2-diethylamino)ethoxy]-m,m-diiodophenyl ketone, 2-n-Butyl-3',5'-diiodo-4'-N-diethylaminoethoxy-3-benzoylbenzofuran, Amidorone, Ancaron, Sedacoron, Sedacorone
IUPAC Name (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone
SMILES CCCCc1oc2ccccc2c1C(=O)c3cc(I)c(OCCN(CC)CC)c(I)c3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C25H29I2NO3/c1-4-7-11-22-23(18-10-8-9-12-21(18)31-22)24(29)17-15-19(26)25(20(27)16-17)30-14-13-28(5-2)6-3/h8-10,12,15-16H,4-7,11,13-14H2,1-3H3
Formula Weight 645.0237
Percent Purity >95%
Chemical Name or Material Amiodarone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.