CAS RN 1951-25-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Amiodarone, TRC
CAS: 1951-25-3 Molecular Formula: C25H29I2NO3 Molecular Weight (g/mol): 645.31 Synonym: 2-butyl-3-benzofuranyl 4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl Ketone,2-Butyl-3-[3,5-diiodo-4-(2-diethylaminoethoxy)benzoyl]benzofuran,2-Butyl-3-benzofuranyl p-[(2-diethylamino)ethoxy]-m,m-diiodophenyl ketone,2-n-Butyl-3',5'-diiodo-4'-N-diethylaminoethoxy-3-benzoylbenzofuran,Amidorone,Ancaron,Sedacoron,Sedacorone IUPAC Name: (2-butyl-1-benzofuran-3-yl)-[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone SMILES: CCCCc1oc2ccccc2c1C(=O)c3cc(I)c(OCCN(CC)CC)c(I)c3