missing translation for 'onlineSavingsMsg'
Learn More

Aminopeptidase A/ENPEP Antibody (6C3), DyLight 488, Novus Biologicals™

Product Code. 18215054 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18215054 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18215054 Supplier Novus Biologicals Supplier No. NBP143281G0.1ML
  • ONLINE EXCLUSIVE PRICE
    563.00 EUR
    532.35 EUR / 0.1mL
    Save 30.65 EUR
    5% Off
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rat Monoclonal Antibody

Aminopeptidase A/ENPEP Monoclonal specifically detects Aminopeptidase A/ENPEP in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aminopeptidase A/ENPEP
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 6C3
Conjugate DyLight 488
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q07075
Gene Alias Aminopeptidase A, APA, AP-A, CD249, CD249 antigen, Differentiation antigen gp160, EAP, EC 3.4.11, EC 3.4.11.7, glutamyl aminopeptidase, glutamyl aminopeptidase (aminopeptidase A), gp160
Gene Symbols ENPEP
Host Species Rat
Immunogen The immunogen for this antibody was Aminopeptidase A/APA.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Cancer, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Neuronal Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2028
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.