missing translation for 'onlineSavingsMsg'
Learn More

Aminoiminomethanesulfinic Acid, Sigma-Aldrich™

Product Code. 15884481
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884481 1 kg 1kg
15874481 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15884481 Supplier Sigma Aldrich Supplier No. 8200821000
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1758-73-2
Grade Research
Molecular Formula HN=C(NH2)SO2H
Molecular Weight (g/mol) 108.12

Specifications

Melting Point 123°C (Decomposition)
Density 1.68 g/cm3 at 20°C
MDL Number MFCD00002397
Quantity 1 kg
Synonym Aminoiminomethanesulfinic acid, FormamidinesulfInic acid, Thiourea dioxide, Aminoiminomethanesulfinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.