accessibility menu, dialog, popup

UserName

H2NNHSOHO

CAS RN 1758-73-2

IUPAC Name: carbamimidoylsulfinic acid
Synonyms: thiourea dioxide formamidine sulfinic acid
Molecular Weight (g/mol): 108.12
Molecular Formula: CH4N2O2S
InChi Key: FYOWZTWVYZOZSI-UHFFFAOYNA-N
SMILES: NC(=N)S(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)