missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AG 1478 hydrochloride
GREENER_CHOICE

Product Code. 30001222
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001222

Brand: Tocris Bioscience™ 1276/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent EGFR-kinase inhibitor

Potent and selective inhibitor of epidermal growth factor receptor kinase (IC50 values 3 nM for EGFR and > 100 μM for ErbB2 and PDGFR). Inhibits proliferation of NCI-H2170 NSCLC cells in vitro (IC50 = 1 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170449-18-0
Molecular Formula C16H15Cl2N3O2

Specifications

Quantity 10 mg
Formula Weight 352.22
Purity >98%
Molecular Weight (g/mol) 352.215
Chemical Name or Material AG 1478 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.