accessibility menu, dialog, popup

UserName

CAS RN 170449-18-0

HClH3CONNNHClOH3C

CAS RN 170449-18-0

IUPAC Name: hydrogen N-(3-chlorophenyl)-6,7-dimethoxyquinazolin-4-amine chloride
Synonyms: hydrogen N-(3-chlorophenyl)-6,7-dimethoxyquinazolin-4-amine chloride
Molecular Weight (g/mol): 352.22
Molecular Formula: C16H15Cl2N3O2
InChi Key: WDJDYIUSDDVWKB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=C(OC)C=C2C(NC3=CC=CC(Cl)=C3)=NC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results