missing translation for 'onlineSavingsMsg'
Learn More

Adipic Acid, Sigma-Aldrich™

Product Code. 15876580
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15876580 100 g 100g
15870531 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15876580 Supplier Sigma Aldrich Supplier No. 8186500100
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-04-9
Molecular Weight (g/mol) 146.14

Specifications

Melting Point 150.9°C
Density 1.36 g/cm3 at 25°C
Quantity 100 g
MDL Number MFCD00004420
Synonym Adipic acid, Hexanedioic acid, Adipic acid
Chemical Name or Material Adipic Acid
compliance-icons
Product Identifier
  • ADIPIC ACID FOR SYNTHESIS
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.