accessibility menu, dialog, popup

UserName

HOOOHO

CAS RN 124-04-9

IUPAC Name: hexanedioic acid
Synonyms: adipic acid acide adipique monoammonium salt adipic acid acifloctin adi-pure fema number 2011 cu(+2) salt adipic acid disodium salt adipic acid adipic acid (substance) adipic acid, mg salt (1:1)
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: WNLRTRBMVRJNCN-UHFFFAOYSA-N
SMILES: OC(=O)CCCCC(O)=O
Molecular Weight (g/mol) 
  • (6)
  • (11)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (7)
  • (3)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (6)
  • (11)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (7)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)