missing translation for 'onlineSavingsMsg'
Learn More

Adenosine A2aR Antibody (7F6-G5-A2), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18214641 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18214641 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18214641 Supplier Novus Biologicals Supplier No. NBP139474AF750
  • ONLINE EXCLUSIVE PRICE
    639.00 EUR
    603.75 EUR / 0.1mL
    Save 35.25 EUR
    5% Off
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Adenosine A2aR Monoclonal specifically detects Adenosine A2aR in Canine, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep samples. It is validated for CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenosine A2aR
Applications CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7F6-G5-A2
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular)
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias adenosine A2 receptor, adenosine A2a receptor, adenosine receptor A2a, adenosine receptor subtype A2a, ADORA 2, ADORA2, hA2aR, RDC8
Gene Symbols ADORA2A
Host Species Mouse
Immunogen Full length recombinant human Adenosine A2a R. [UniProtNo. P29274]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 135
Target Species Canine, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.