missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-diphosphate Monosodium Salt, 97.7%, MP Biomedicals™

Product Code. 11454592 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454592 1 g 1g
11444592 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454592 Supplier MP Biomedicals Supplier No. 0219014901
  • ONLINE EXCLUSIVE PRICE
    97.00 EUR
    92.40 EUR / 1g
    Save 4.60 EUR
    4% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 20398-34-9
Molecular Formula C10H15N5O10P2
Molecular Weight (g/mol) 427.20
MDL Number MFCD00066473,MFCD00065470,MFCD00066635
InChI Key XTWYTFMLZFPYCI-KQYNXXCUSA-N
Synonym adp monosodium salt, adenosine 5'-diphosphate, sodium salt, adenosine 5'-diphosphate sodium salt
PubChem CID 87704518
IUPAC Name [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid
SMILES NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Assay Percent Range 0.977
Quantity 1 g
Formula Weight 449.19
Physical Form Powder
Chemical Name or Material Adenosine-5'-diphosphate Monosodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.