accessibility menu, dialog, popup

UserName

CAS RN 20398-34-9

IUPAC Name: [({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy]phosphonic acid
Synonyms: adenosine-diphosphate magnesium adp MgADP adenosine 5' pyrophosphate ADP pyrophosphate, adenosine adp adenosine-5-diphosphate 5'-adenylphosphoric acid adenosine diphosphate (substance)
Molecular Weight (g/mol): 427.2
Molecular Formula: C10H15N5O10P2
InChi Key: XTWYTFMLZFPYCI-KQYNXXCUSA-N
SMILES: NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results