missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine-5'-diphosphate Monosodium Salt, 97.7%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
Specifications
Specifications
| Color | White |
| Assay Percent Range | 0.977 |
| Quantity | 500 mg |
| Formula Weight | 449.19 |
| Physical Form | Powder |
| Chemical Name or Material | Adenosine-5'-diphosphate Monosodium Salt |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction