missing translation for 'onlineSavingsMsg'
Learn More

Acrylamide, TRC

Product Code. 30134362
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134362 25 g 25g
30040924 1 g 1g
30040395 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30134362 Supplier Toronto Research Chemicals Supplier No. TRCA19130025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-06-1
Molecular Formula C3 H5 N O
Molecular Weight (g/mol) 71.08
Synonym Acrylamide (8CI), Acrylic amide, Bio-Acrylamide 50, DM 206, Ethylenecarboxamide, NSC 7785, Propenamide, Vinyl amide
IUPAC Name prop-2-enamide
SMILES NC(=O)C=C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)
Formula Weight 71.0371
Chemical Name or Material Acrylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.