CAS RN 79-06-1
Thermo Scientific Chemicals Acrylamide / N,N'-Methylenebisacrylamide 29:1, for biochemistry, 40% mix solution in water
Acrylamide / N,N'-Methylenebisacrylamide, CAS # 79-06-1, 7732-18-5, 110-26-9, 79-06-1, is a water soluble compound that cross-links to produce polyacrylamide gels that are widely used in chromatography and the electrophoretic separation of nucleic acids and proteins. | CAS: 79-06-1 | C3H5NO | 71.08 g/mol
Acrylamide: Bis-Acrylamide 29:1 (40% Solution/Electrophoresis), Fisher BioReagents
CAS: 79-06-1 | C3H5NO | 71.08 g/mol
Thermo Scientific Chemicals Acrylamide / N,N'-Methylenebisacrylamide 37.5:1, for biochemistry, 40% mix solution in water
Acrylamide / N,N'-Methylenebisacrylamide, CAS # 79-06-1, 7732-18-5, 110-26-9, 79-06-1, is a water soluble compound that cross-links to produce polyacrylamide gels that are widely used in chromatography and the electrophoretic separation of nucleic acids and proteins. | CAS: 79-06-1 | C3H5NO | 71.08 g/mol
Thermo Scientific Chemicals Acrylamide (40%) Solutions
CAS: 79-06-1 Molecular Formula: C3H5NO Molecular Weight (g/mol): 71.08 MDL Number: MFCD00008032 InChI Key: HRPVXLWXLXDGHG-UHFFFAOYSA-N PubChem CID: 6579 ChEBI: CHEBI:28619 IUPAC Name: prop-2-enamide SMILES: NC(=O)C=C
Thermo Scientific Chemicals Acrylamide, 98.5%, extra pure
CAS: 79-06-1 Molecular Formula: C3H5NO Molecular Weight (g/mol): 71.08 MDL Number: MFCD00008032 InChI Key: HRPVXLWXLXDGHG-UHFFFAOYSA-N Synonym: Ethylenecarboxamide,2-Propenamide PubChem CID: 6579 ChEBI: CHEBI:28619 IUPAC Name: prop-2-enamide SMILES: NC(=O)C=C
Thermo Scientific Chemicals Acrylamide (30%) Solutions
CAS: 79-06-1 Molecular Formula: C3H5NO Molecular Weight (g/mol): 71.08 MDL Number: MFCD00008032 InChI Key: HRPVXLWXLXDGHG-UHFFFAOYSA-N PubChem CID: 6579 ChEBI: CHEBI:28619 IUPAC Name: prop-2-enamide SMILES: NC(=O)C=C
Thermo Scientific Chemicals Acrylamide, 99+%, for biochemistry, electrophoresis grade
CAS: 79-06-1 Molecular Formula: C3H5NO Molecular Weight (g/mol): 71.08 MDL Number: MFCD00008032 InChI Key: HRPVXLWXLXDGHG-UHFFFAOYSA-N Synonym: Ethylenecarboxamide,2-Propenamide PubChem CID: 6579 ChEBI: CHEBI:28619 IUPAC Name: prop-2-enamide SMILES: NC(=O)C=C
Acrylamide, 99.9%
CAS: 79-06-1 Molecular Formula: C3H5NO Molecular Weight (g/mol): 71.08 MDL Number: MFCD00008032 InChI Key: HRPVXLWXLXDGHG-UHFFFAOYSA-N Synonym: acrylamide,2-propenamide,propenamide,acrylic amide,akrylamid,ethylenecarboxamide,vinyl amide,acrylic acid amide,2-propeneamide,propeneamide PubChem CID: 6579 ChEBI: CHEBI:28619 IUPAC Name: prop-2-enamide SMILES: NC(=O)C=C
Thermo Scientific™ Acrylamide, electrophoresis grade, 99+%
CAS: 79-06-1 Molecular Formula: C3H5NO Molecular Weight (g/mol): 71.08 MDL Number: MFCD00008032 InChI Key: HRPVXLWXLXDGHG-UHFFFAOYSA-N Synonym: acrylamide,2-propenamide,propenamide,acrylic amide,akrylamid,ethylenecarboxamide,vinyl amide,acrylic acid amide,2-propeneamide,propeneamide PubChem CID: 6579 ChEBI: CHEBI:28619 IUPAC Name: prop-2-enamide SMILES: NC(=O)C=C
Thermo Scientific Chemicals Acrylamide / N,N'-Methylenebisacrylamide 19:1, for biochemistry, 40% mix solution in water
CAS: 79-06-1 | C3H5NO | 71.08 g/mol
Thermo Scientific Chemicals Acrylamide, 99.5%, Molecular Biology Grade
CAS: 79-06-1 Molecular Formula: C3H5NO Molecular Weight (g/mol): 71.08 MDL Number: MFCD00008032 InChI Key: HRPVXLWXLXDGHG-UHFFFAOYSA-N Synonym: acrylamide,2-propenamide,propenamide,acrylic amide,akrylamid,ethylenecarboxamide,vinyl amide,acrylic acid amide,2-propeneamide,propeneamide PubChem CID: 6579 ChEBI: CHEBI:28619 IUPAC Name: prop-2-enamide SMILES: NC(=O)C=C
Acrylamide in Methanol 100μg/mL, Fisher Chemical™
Acrylamide, 98+%
CAS: 79-06-1 Molecular Formula: C3H5NO Molecular Weight (g/mol): 71.08 MDL Number: MFCD00008032 InChI Key: HRPVXLWXLXDGHG-UHFFFAOYSA-N Synonym: acrylamide,2-propenamide,propenamide,acrylic amide,akrylamid,ethylenecarboxamide,vinyl amide,acrylic acid amide,2-propeneamide,propeneamide PubChem CID: 6579 ChEBI: CHEBI:28619 IUPAC Name: prop-2-enamide SMILES: NC(=O)C=C