missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Acrosin Monoclonal specifically detects Acrosin in Bovine (Negative), Canine (Negative), Human (Negative), Pig samples. It is validated for CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Western Blot.
Specifications
Specifications
| Antigen | Acrosin |
| Applications | CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Western Blot |
| Classification | Monoclonal |
| Clone | ACR-2 |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | acrosin, ACRS, preproacrosin, proacrosin |
| Gene Symbols | ACR |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?