missing translation for 'onlineSavingsMsg'
Learn More

Acamprosate Calcium, TRC

Product Code. 30137226
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137226 100 mg 100mg
30133654 1 g 1g
30004129 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137226 Supplier Toronto Research Chemicals Supplier No. TRCA120000100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77337-73-6
Molecular Formula 2 C5 H10 N O4 S . Ca
Molecular Weight (g/mol) 400.48
Synonym Acamprosate calcium, Calcium bis[3-(acetylamino)propane-1-sulfonate], 1-Propanesulfonic acid, 3-(acetylamino)-, calcium salt (2:1), Alcomed, Aotal, Ca-AOTA, Calcium N-acetylhomotaurinate, Calcium acamprosate, Calcium bisacetyl homotaurine, Campral EC, Regtect, Sobriol
IUPAC Name calcium;3-acetamidopropane-1-sulfonate
SMILES [Ca+2].CC(=O)NCCCS(=O)(=O)[O-].CC(=O)NCCCS(=O)(=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/2C5H11NO4S.Ca/c2*1-5(7)6-3-2-4-11(8,9)10;/h2*2-4H2,1H3,(H,6,7)(H,8,9,10);/q;;+2/p-2
Formula Weight 400.0287
Chemical Name or Material Acamprosate Calcium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.