accessibility menu, dialog, popup

UserName

CAS RN 77337-73-6

CaH3COHNSOOOH3COHNSOOO

CAS RN 77337-73-6

IUPAC Name: calcium bis(3-acetamidopropane-1-sulfonate)
Synonyms: acamprosate calcium
Molecular Weight (g/mol): 400.47
Molecular Formula: C10H20CaN2O8S2
InChi Key: BUVGWDNTAWHSKI-UHFFFAOYSA-L
SMILES: [Ca++].CC(=O)NCCCS([O-])(=O)=O.CC(=O)NCCCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)