missing translation for 'onlineSavingsMsg'
Learn More

AACOCF3, TRC

Product Code. 30179485
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179485 5 mg 5mg
30179635 1 mg 1mg
30179105 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179485 Supplier Toronto Research Chemicals Supplier No. TRCA1045085MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149301-79-1
Molecular Formula C21H31F3O
Molecular Weight (g/mol) 356.47
Synonym (all-Z)-1,1,1-Trifluoro-6,9,12,15-heneicosatetraen-2-one, (6Z,9Z,12Z,15Z)-1,1,1-Trifluoro-6,9,12,15-heneicosatetraen-2-one, AN 20579, ATFMK, Arachidonyl Trifluoromethyl Ketone, BM 162353, L 734575, (all-Z)-1,1-Trifluoro-, PACOCF31, AACOCF3
SMILES CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)C(F)(F)F

Specifications

Quantity 5 mg
Chemical Name or Material 1,1,1-Trifluoro-6Z,9Z,12Z,15Z-heneicosateraen-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.