CAS RN 149301-79-1
IUPAC Name:
(6Z,9Z,12Z,15Z)-1,1,1-trifluorohenicosa-6,9,12,15-tetraen-2-one
Synonyms:
AACOCF3 arachidonic acid trifluoromethyl ketone aacocf3 arachidonyl trifluoromethyl ketone arachidonyltrifluoromethane
Molecular Weight (g/mol):
356.47
Molecular Formula:
C21H31F3O
InChi Key:
PLWROONZUDKYKG-DOFZRALJSA-N
SMILES:
CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)C(F)(F)F
1
–
2
of
2
results
1
AACOCF3, TRC
CAS: 149301-79-1 Molecular Formula: C21H31F3O Molecular Weight (g/mol): 356.47 Synonym: (all-Z)-1,1,1-Trifluoro-6,9,12,15-heneicosatetraen-2-one,(6Z,9Z,12Z,15Z)-1,1,1-Trifluoro-6,9,12,15-heneicosatetraen-2-one,AN 20579,ATFMK,Arachidonyl Trifluoromethyl Ketone,BM 162353,L 734575,(all-Z)-1,1-Trifluoro-,PACOCF31,AACOCF3 SMILES: CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)C(F)(F)F
2
Tocris Bioscience™ AACOCF3
CAS: 149301-79-1 Molecular Formula: C21H31F3O