missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxybentazone, TRC

Product Code. 17911855
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17911855 1 mg 1mg
17911315 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17911855 Supplier Toronto Research Chemicals Supplier No. TRCH8065051MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60374-43-8
Molecular Formula C10 H12 N2 O4 S
Molecular Weight (g/mol) 256.27828
Synonym 8-Hydroxybentazon, 8-Hydroxybentazone
IUPAC Name 8-hydroxy-2,2-dioxo-3-propan-2-yl-1H-2λ^{6},1,3-benzothiadiazin-4-one
SMILES CC(C)N1C(=O)c2cccc(O)c2NS1(=O)=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C10H12N2O4S/c1-6(2)12-10(14)7-4-3-5-8(13)9(7)11-17(12,15)16/h3-6,11,13H,1-2H3
Formula Weight 256.0518
Percent Purity >95%
Chemical Name or Material Bentazone-8-hydroxy
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.