CAS RN 60374-43-8
CAS RN 60374-43-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
8-Hydroxybentazone, TRC
CAS: 60374-43-8 Molecular Formula: C10 H12 N2 O4 S Molecular Weight (g/mol): 256.27828 Synonym: 8-Hydroxybentazon,8-Hydroxybentazone IUPAC Name: 8-hydroxy-2,2-dioxo-3-propan-2-yl-1H-2λ^{6},1,3-benzothiadiazin-4-one SMILES: CC(C)N1C(=O)c2cccc(O)c2NS1(=O)=O