Learn More
7-Desmethylagomelatine, TRC
Product Code. 30360511
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360511 10 mg 10mg
30360812 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30360511 Supplier Toronto Research Chemicals Supplier No. TRCD23109510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 152302-45-9
Molecular Formula C14 H15 N O2
Molecular Weight (g/mol) 229.27
Synonym Acetamide, N-[2-(7-hydroxy-1-naphthalenyl)ethyl]-, N-[2-(7-Hydroxy-1-naphthalenyl)ethyl]acetamide, 7-Desmethylagomelatine, N-[2-(7-Hydroxy-1-naphthyl)ethyl]acetamide, O-Demethylagomelatine, S 21517
IUPAC Name N-[2-(7-hydroxynaphthalen-1-yl)ethyl]acetamide
SMILES CC(=O)NCCc1cccc2ccc(O)cc12
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C14H15NO2/c1-10(16)15-8-7-12-4-2-3-11-5-6-13(17)9-14(11)12/h2-6,9,17H,7-8H2,1H3,(H,15,16)
Formula Weight 229.1103
Percent Purity >95%
Chemical Name or Material 7-Desmethyl Agomelatine
Grade HPLC