CAS RN 152302-45-9
CAS RN 152302-45-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Desmethylagomelatine, TRC
CAS: 152302-45-9 Molecular Formula: C14 H15 N O2 Molecular Weight (g/mol): 229.27 Synonym: Acetamide, N-[2-(7-hydroxy-1-naphthalenyl)ethyl]-,N-[2-(7-Hydroxy-1-naphthalenyl)ethyl]acetamide,7-Desmethylagomelatine,N-[2-(7-Hydroxy-1-naphthyl)ethyl]acetamide,O-Demethylagomelatine,S 21517 IUPAC Name: N-[2-(7-hydroxynaphthalen-1-yl)ethyl]acetamide SMILES: CC(=O)NCCc1cccc2ccc(O)cc12