missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy Chlorzoxazone, TRC

Product Code. 30004390
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004390 50 mg 50mg
30005634 10 mg 10mg
30003756 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004390 Supplier Toronto Research Chemicals Supplier No. TRCH82512050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1750-45-4
Molecular Formula C7 H4 Cl N O3
Molecular Weight (g/mol) 185.56
Synonym 2(3H)-Benzoxazolone, 5-chloro-6-hydroxy-, 2,6-Benzoxazolediol, 5-chloro- (6CI), 2-Benzoxazolinone, 5-chloro-6-hydroxy- (7CI,8CI), 6-Hydroxychloroxazone, 6-Hydroxychlorzoxazone, 6-Hydroxylchlorzoxazone, NSC 24955
IUPAC Name 5-chloro-6-hydroxy-3H-1,3-benzoxazol-2-one
SMILES Oc1cc2OC(=O)Nc2cc1Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H4ClNO3/c8-3-1-4-6(2-5(3)10)12-7(11)9-4/h1-2,10H,(H,9,11)
Formula Weight 184.988
Percent Purity >95%
Chemical Name or Material 6-Hydroxy Chlorzoxazone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.