CAS RN 1750-45-4
CAS RN 1750-45-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Hydroxy Chlorzoxazone, TRC
CAS: 1750-45-4 Molecular Formula: C7 H4 Cl N O3 Molecular Weight (g/mol): 185.56 Synonym: 2(3H)-Benzoxazolone, 5-chloro-6-hydroxy-,2,6-Benzoxazolediol, 5-chloro- (6CI),2-Benzoxazolinone, 5-chloro-6-hydroxy- (7CI,8CI),6-Hydroxychloroxazone,6-Hydroxychlorzoxazone,6-Hydroxylchlorzoxazone,NSC 24955 IUPAC Name: 5-chloro-6-hydroxy-3H-1,3-benzoxazol-2-one SMILES: Oc1cc2OC(=O)Nc2cc1Cl