Learn More
6-Chloro-2-fluoro-3-methoxyaniline, 97%, Thermo Scientific™
Product Code. 15424417
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15424417 1 g 1g
15434417 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15424417 Supplier Thermo Scientific Alfa Aesar Supplier No. H31779.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
6-Chloro-2-fluoro-3-methoxyaniline is used as a protected form of ammonia in a practical synthesis of azetidine
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1017777-77-3
Molecular Formula C7H7ClFNO
Molecular Weight (g/mol) 175.587
MDL Number MFCD09832271
InChI Key ABRCLZVZMHSIRQ-UHFFFAOYSA-N
Synonym 6-chloro-2-fluoro-3-methoxyphenylamine
PubChem CID 46737508
IUPAC Name 6-chloro-2-fluoro-3-methoxyaniline
SMILES COC1=C(C(=C(C=C1)Cl)N)F
Specifications
Melting Point 40°C to 43°C
Quantity 1 g
UN Number UN2811
Formula Weight 175.59
Percent Purity 97%
Chemical Name or Material 6-Chloro-2-fluoro-3-methoxyaniline