accessibility menu, dialog, popup

UserName

H3COClNH2F

CAS RN 1017777-77-3

IUPAC Name: 6-chloro-2-fluoro-3-methoxyaniline
Synonyms: 6-chloro-2-fluoro-3-methoxyaniline
Molecular Weight (g/mol): 175.59
Molecular Formula: C7H7ClFNO
InChi Key: ABRCLZVZMHSIRQ-UHFFFAOYSA-N
SMILES: COC1=C(F)C(N)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results