Learn More
6-Chloro-2,3-difluorobenzoyl chloride, 97%, Thermo Scientific™
Product Code. 15426767
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426767 5 g 5g
15416767 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15426767 Supplier Thermo Scientific Alfa Aesar Supplier No. H33875.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1208078-25-4
Molecular Formula C7H2Cl2F2O
Molecular Weight (g/mol) 210.989
MDL Number MFCD12922578
InChI Key CHLDUAMDKJNJKC-UHFFFAOYSA-N
PubChem CID 50998364
IUPAC Name 6-chloro-2,3-difluorobenzoyl chloride
SMILES C1=CC(=C(C(=C1F)F)C(=O)Cl)Cl
Specifications
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 211
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 6-Chloro-2,3-difluorobenzoyl chloride