accessibility menu, dialog, popup

UserName

FFOClCl

CAS RN 1208078-25-4

IUPAC Name: 6-chloro-2,3-difluorobenzoyl chloride
Synonyms: 6-chloro-2,3-difluorobenzoyl chloride
Molecular Weight (g/mol): 210.99
Molecular Formula: C7H2Cl2F2O
InChi Key: CHLDUAMDKJNJKC-UHFFFAOYSA-N
SMILES: FC1=C(F)C(C(Cl)=O)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results