missing translation for 'onlineSavingsMsg'
Learn More

6-Aminoquinoline-5-carbonitrile, 95%, Thermo Scientific™

Product Code. 15425248
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425248 1 g 1g
15435248 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425248 Supplier Thermo Scientific Alfa Aesar Supplier No. H59061.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 54398-51-5
Molecular Formula C10H7N3
Molecular Weight (g/mol) 169.187
MDL Number MFCD05855932
InChI Key PYCIBXGKTGBHHF-UHFFFAOYSA-N
Synonym 6-amino-5-quinolinecarbonitrile, 6-amino-quinoline-5-carbonitrile, 5-quinolinecarbonitrile, 6-amino, pycibxgktgbhhf-uhfffaoysa, quinoline-5-carbonitrile, 6-amino
PubChem CID 1507240
IUPAC Name 6-aminoquinoline-5-carbonitrile
SMILES C1=CC2=C(C=CC(=C2C#N)N)N=C1

Tekniske data

Quantity 1 g
Formula Weight 169.18
Percent Purity 95%
Chemical Name or Material 6-Aminoquinoline-5-carbonitrile
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.