accessibility menu, dialog, popup

UserName

CAS RN 54398-51-5

IUPAC Name: 6-aminoquinoline-5-carbonitrile
Synonyms: 6-aminoquinoline-5-carbonitrile
Molecular Weight (g/mol): 169.19
Molecular Formula: C10H7N3
InChi Key: PYCIBXGKTGBHHF-UHFFFAOYSA-N
SMILES: NC1=C(C#N)C2=CC=CN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results