Learn More
6,9-Dichloro-2-methoxyacridine, 98%
Product Code. 11307645
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307645 5 g 5g
11317645 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11307645 Supplier Thermo Scientific Alfa Aesar Supplier No. L05913.06
Request Bulk or Custom Format
CAS: 86-38-4 | C14H9Cl2NO | 278.132 g/mol
6,9-Dichloro-2-methoxyacridine is used to prepare 6-chloro-2-methoxy-acridin-9-ylamine by reacting with phenol and ammonium carbonate. It is used in the preparation of 9-amino-6-chloro-2-methoxyacridine, N'-(6-Chloro-2-methoxy-acridin-9-yl)-heptylamine and N,N'-bis-(6-chloro-2-methoxy-acridin-9-yl)-hexane-1,6-diamine. Further, it is involved in the preparation of 4-aminoquinoline and 9-aminoacridine derivatives by reaction with quinolizidinylalkylamines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6,9-Dichloro-2-methoxyacridine is used to prepare 6-chloro-2-methoxy-acridin-9-ylamine by reacting with phenol and ammonium carbonate. It is used in the preparation of 9-amino-6-chloro-2-methoxyacridine, N′-(6-Chloro-2-methoxy-acridin-9-yl)-heptylamine and N,N′-bis-(6-chloro-2-methoxy-acridin-9-yl)-hexane-1,6-diamine. Further, it is involved in the preparation of 4-aminoquinoline and 9-aminoacridine derivatives by reaction with quinolizidinylalkylamines.
Solubility
Insoluble in water.
Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 86-38-4
Molecular Formula C14H9Cl2NO
Molecular Weight (g/mol) 278.132
MDL Number MFCD00005028
InChI Key RYRNQWYNHLLOGX-UHFFFAOYSA-N
Synonym halocrin, 3,9-dichloro-7-methoxyacridine, halocrine, 6.9-dichloro-2-methoxyacridine, 2-methoxy-6,9-dichloroacridine, acridine,9-dichloro-2-methoxy, acridine, 6,9-dichloro-2-methoxy, bidd:gt0206, 6,9dichloro-2-methoxyacridine, 6,9-dichloro-2-methoxy-acridine
PubChem CID 66577
IUPAC Name 6,9-dichloro-2-methoxyacridine
SMILES COC1=CC2=C(C3=C(C=C(C=C3)Cl)N=C2C=C1)Cl
Specifications
Melting Point 163°C to 165°C
Quantity 5 g
Beilstein 226309
Solubility Information Insoluble in water.
Formula Weight 278.14
Percent Purity 98%
Chemical Name or Material 6,9-Dichloro-2-methoxyacridine