CAS RN 86-38-4
CAS RN 86-38-4
IUPAC Name:
6,9-dichloro-2-methoxyacridine
Synonyms:
6,9-dichloro-2-methoxyacridine
Molecular Weight (g/mol):
278.13
Molecular Formula:
C14H9Cl2NO
InChi Key:
RYRNQWYNHLLOGX-UHFFFAOYSA-N
SMILES:
COC1=CC=C2N=C3C=C(Cl)C=CC3=C(Cl)C2=C1
1
–
1
of
1
results
1
6,9-Dichloro-2-methoxyacridine, 98%
CAS: 86-38-4 Molecular Formula: C14H9Cl2NO Molecular Weight (g/mol): 278.132 MDL Number: MFCD00005028 InChI Key: RYRNQWYNHLLOGX-UHFFFAOYSA-N Synonym: halocrin,3,9-dichloro-7-methoxyacridine,halocrine,6.9-dichloro-2-methoxyacridine,2-methoxy-6,9-dichloroacridine,acridine,9-dichloro-2-methoxy,acridine, 6,9-dichloro-2-methoxy,bidd:gt0206,6,9dichloro-2-methoxyacridine,6,9-dichloro-2-methoxy-acridine PubChem CID: 66577 IUPAC Name: 6,9-dichloro-2-methoxyacridine SMILES: COC1=CC2=C(C3=C(C=C(C=C3)Cl)N=C2C=C1)Cl