Learn More
Invitrogen™ 5X Kinase Buffer A
Description
The 5X Kinase Buffer A is a concentrated general kinase assay buffer that can be used with a variety of assay technologies such as, LanthaScreen™, Adapta™, and Z’-LYTE™ assays.
Ready to use and validated with LanthaScreen™, Adapta™, and Z’LYTE™ assays
Optimized 5X concentrated formulation
Simplified handling with room temperature storage
Easily Screen for Kinase Targets using an Invitrogen™ Platform
For more information on the kinase assays that can be used with our 5X Kinase Buffer A see the following links:
LanthaScreen™ Eu Binding Assay
LanthaScreen™ Activity Assay
Adapta™ Universal Kinase Assay
Z´-LYTE™ Kinase Assay Kits
Save Freezer Space
The 5X Kinase Buffer can be stored at room temperature, saving you time and precious freezer space.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Ready to use and validated with LanthaScreen™, Adapta™, and Z’LYTE™ assays
Easily Screen for Kinase Targets using an Invitrogen™ Platform
For more information on the kinase assays that can be used with our 5X Kinase Buffer A see the following links:
LanthaScreen™ Eu Binding Assay
LanthaScreen™ Activity Assay
Adapta™ Universal Kinase Assay
Z´-LYTE™ Kinase Assay Kits
Save Freezer Space
The 5X Kinase Buffer can be stored at room temperature, saving you time and precious freezer space.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Specifications
Specifications
| Chemical Name or Material | Kinase Buffer A |
| Concentration | 5X |
| For Use With (Application) | Pharma & Biopharma; Protein Assays and Analysis |
| For Use With (Equipment) | Adapta™ and Z-LYTE™ assays; |
| pH | 7.5 |
| Physical Form | Liquid |
| Quantity | 100 ml |
| Includes | 250 mM HEPES (pH 7.5), 50 mM MgCl2, 5 mM EGTA, and 0.05% Brij-35 |