missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-DL-serine, TRC

Product Code. 30240661
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240661 50 mg 50mg
30240591 100 mg 100mg
30240935 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240661 Supplier Toronto Research Chemicals Supplier No. TRCG65011050MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 687-38-7
Molecular Formula C5H10N2O4
Molecular Weight (g/mol) 162.14
Synonym Glycylserine, 2-(2-Aminoacetamido)-3-hydroxypropanoic Acid, Glycyl-DL-serine, NSC 163326
IUPAC Name 2-[(2-aminoacetyl)amino]-3-hydroxypropanoic acid
SMILES NCC(=O)NC(CO)C(=O)O

Specifications

InChI Formula InChI=1S/C5H10N2O4/c6-1-4(9)7-3(2-8)5(10)11/h3,8H,1-2,6H2,(H,7,9)(H,10,11)
Quantity 50 mg
Formula Weight 162.0641
Shipping Condition Room Temperature
Chemical Name or Material Glycyl-DL-serine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.