CAS RN 687-38-7
IUPAC Name:
2-(2-aminoacetamido)-3-hydroxypropanoic acid
Synonyms:
2-(2-aminoacetamido)-3-hydroxypropanoic acid
Molecular Weight (g/mol):
162.15
Molecular Formula:
C5H10N2O4
InChi Key:
BCCRXDTUTZHDEU-UHFFFAOYNA-N
SMILES:
NCC(=O)NC(CO)C(O)=O
1
–
1
of
1
results
1
Glycyl-DL-serine, TRC
CAS: 687-38-7 Molecular Formula: C5H10N2O4 Molecular Weight (g/mol): 162.14 Synonym: Glycylserine,2-(2-Aminoacetamido)-3-hydroxypropanoic Acid,Glycyl-DL-serine,NSC 163326 IUPAC Name: 2-[(2-aminoacetyl)amino]-3-hydroxypropanoic acid SMILES: NCC(=O)NC(CO)C(=O)O