missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfosalicylic Acid Dihydrate, Sigma-Aldrich™

Product Code. 15896250
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896250 1 kg 1kg
15886250 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15896250 Supplier Sigma Aldrich Supplier No. 8006911000
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5965-83-3
Grade Research
Molecular Weight (g/mol) 254.21

Specifications

Melting Point 109°C to 117°C (Anhydrous Substance)
Density 0.8 g/cm3 at 20°C
MDL Number MFCD00149540
Quantity 1 kg
Synonym 5-Sulfosalicylic acid dihydrate, Salicylsulfonic acid, 5-Sulfosalicylic acid dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.