accessibility menu, dialog, popup

UserName

HOOHOSOHOOH2OH2O

CAS RN 5965-83-3

IUPAC Name: 2-hydroxy-5-sulfobenzoic acid; bis(methane)
Synonyms: bis(methane); sulfosalicylic acid
Molecular Weight (g/mol): 250.27
Molecular Formula: C9H14O6S
InChi Key: NFYHZVWMQHQKRU-UHFFFAOYSA-N
SMILES: C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
Molecular Weight (g/mol) 
  • (16)
  • (6)
Quantity 
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (6)
  • (3)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
Grade 
  • (3)
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (8)
  • (6)
  • (3)
  • (16)
  • (6)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (6)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)