missing translation for 'onlineSavingsMsg'
Learn More

5-Pentadecylresorcinol, TRC

Product Code. 30133831
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133831 50 mg 50mg
30011608 10 mg 10mg
30012450 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133831 Supplier Toronto Research Chemicals Supplier No. TRCP26700050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3158-56-3
Molecular Formula C21H36O2
Molecular Weight (g/mol) 320.51
Synonym 5-Pentadecyl-1,3-benzenediol, 3-n-Pentadecyl-5-hydroxyphenol, 5-(n-Pentadecyl)resorcinol, Adipostatin A, Hydrogenated Cardol, NSC 776
IUPAC Name 5-pentadecylbenzene-1,3-diol
SMILES O=C(Nc1cc(c(Cl)cc1)Oc2c(Cl)cc(cn2)C(F)(F)F)NC(=O)c3c(F)cccc3F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19-16-20(22)18-21(23)17-19/h16-18,22-23H,2-15H2,1H3
Formula Weight 320.2715
Percent Purity >95%
Chemical Name or Material 5-Pentadecylresorcinol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.